Assay Details
Gene Symbol: ripor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012499.2 | NP_001012517.1 | 11 - 12 | 1062 | 71 | |
XM_005166267.4 | XP_005166324.1 | 11 - 12 | 1250 | 71 | ||
XM_009302697.3 | XP_009300972.1 | 11 - 12 | 1216 | 71 | ||
GenBank mRNA | BC090162.1 | - | 11 - 12 | 1066 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
