Assay Details
Gene Symbol: ripor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012499.2 | NP_001012517.1 | 4 - 5 | 454 | 130 | |
XM_005166267.4 | XP_005166324.1 | 4 - 5 | 642 | 130 | ||
XM_009302697.3 | XP_009300972.1 | 4 - 5 | 608 | 130 | ||
GenBank mRNA | BC090162.1 | - | 4 - 5 | 458 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
