Assay Details
Gene Symbol: nmnat1-rbp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017629.2 | NP_001017629.1 | 4 - 5 | 462 | 126 | |
GenBank mRNA | BC092821.1 | - | 4 - 5 | 464 | 126 | |
BC165446.1 | - | 3 - 4 | 454 | 126 | ||
CO804002.1 | - | 4 - 5 | 472 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
