Assay Details
Gene Symbol: hddc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017835.1 | NP_001017835.1 | 1 - 2 | 319 | 110 | |
GenBank mRNA | BC092780.1 | - | 1 - 2 | 319 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
