Assay Details
Gene Symbol: amacr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020626.1 | NP_001018462.1 | 5 - 6 | 830 | 98 | |
GenBank mRNA | BC095377.1 | - | 5 - 6 | 830 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
