Assay Details
Gene Symbol: usta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032729.1 | NP_001027901.1 | 1 - 2 | 343 | 64 | |
XM_009294731.3 | XP_009293006.1 | 1 - 2 | 1532 | 64 | ||
GenBank mRNA | AB205103.1 | - | 1 - 2 | 343 | 64 | |
BC171379.1 | - | 1 - 2 | 332 | 64 | ||
BC171381.1 | - | 1 - 2 | 332 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
