Assay Details
Gene Symbol: usta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032729.1 | NP_001027901.1 | 2 - 3 | 387 | 79 | |
XM_009294731.3 | XP_009293006.1 | 2 - 3 | 1576 | 79 | ||
GenBank mRNA | AB205103.1 | - | 2 - 3 | 387 | 79 | |
BC171379.1 | - | 2 - 3 | 376 | 79 | ||
BC171381.1 | - | 2 - 3 | 376 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
