Assay Details
Gene Symbol: znf598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033718.1 | NP_001028890.1 | 3 - 4 | 530 | 68 | |
GenBank mRNA | BC057520.1 | - | 3 - 4 | 560 | 68 | |
BC171438.1 | - | 3 - 4 | 499 | 68 | ||
BC171442.1 | - | 3 - 4 | 499 | 68 | ||
DQ173495.1 | - | 3 - 4 | 530 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
