Assay Details
Gene Symbol: vps51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042735.1 | NP_001036200.1 | 2 - 3 | 309 | 119 | |
GenBank mRNA | BC162801.1 | - | 2 - 3 | 409 | 119 | |
BC162802.1 | - | 2 - 3 | 409 | 119 | ||
DQ641948.1 | - | 2 - 3 | 309 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
