Assay Details
Gene Symbol: vps51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042735.1 | NP_001036200.1 | 8 - 9 | 2009 | 85 | |
GenBank mRNA | BC162801.1 | - | 8 - 9 | 2109 | 85 | |
BC162802.1 | - | 8 - 9 | 2109 | 85 | ||
DQ641948.1 | - | 8 - 9 | 2009 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
