Assay Details
Gene Symbol: kctd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045434.1 | NP_001038899.1 | 5 - 6 | 1022 | 79 | |
GenBank mRNA | BC122218.1 | - | 5 - 6 | 1022 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
