Assay Details
Gene Symbol: phyhipla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080088.1 | NP_001073557.1 | 1 - 2 | 547 | 61 | |
XM_005158677.4 | XP_005158734.1 | 2 - 3 | 569 | 61 | ||
XM_009292975.3 | XP_009291250.1 | 2 - 3 | 566 | 61 | ||
GenBank mRNA | BC128850.1 | - | 1 - 2 | 547 | 61 | |
BC153387.1 | - | 2 - 3 | 323 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
