Assay Details
Gene Symbol: phyhipla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080088.1 | NP_001073557.1 | 4 - 5 | 1026 | 80 | |
XM_005158677.4 | XP_005158734.1 | 5 - 6 | 1048 | 80 | ||
XM_009292975.3 | XP_009291250.1 | 5 - 6 | 1045 | 80 | ||
GenBank mRNA | BC128850.1 | - | 4 - 5 | 1026 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
