Assay Details
Gene Symbol: rcc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082803.1 | NP_001076272.1 | 4 - 5 | 744 | 71 | |
GenBank mRNA | BC133897.1 | - | 4 - 5 | 744 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
