Assay Details
Gene Symbol: hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102593.1 | NP_001096063.1 | 2 - 3 | 198 | 63 | |
XM_005169908.4 | XP_005169965.1 | 2 - 3 | 558 | 63 | ||
GenBank mRNA | EF150846.1 | - | 2 - 3 | 198 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
