Assay Details
Gene Symbol: golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105599.1 | NP_001099069.1 | 3 - 4 | 329 | 80 | |
XM_021479212.1 | XP_021334887.1 | 3 - 4 | 379 | 80 | ||
GenBank mRNA | BC071425.1 | - | 3 - 4 | 328 | 80 | |
BC153595.1 | - | 3 - 4 | 329 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
