Assay Details
Gene Symbol: psmb13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131677.2 | NP_571752.2 | 1 - 2 | 105 | 78 | |
GenBank mRNA | AF155577.1 | - | 1 - 2 | 78 | ||
BC092899.1 | - | 1 - 2 | 78 | |||
BC165548.1 | - | 1 - 2 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
