Assay Details
Gene Symbol: psmb13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131677.2 | NP_571752.2 | 4 - 5 | 437 | 125 | |
GenBank mRNA | AF155577.1 | - | 4 - 5 | 125 | ||
BC092899.1 | - | 4 - 5 | 125 | |||
BC165548.1 | - | 4 - 5 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
