Assay Details
Gene Symbol: dohh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199563.2 | NP_955857.1 | 2 - 3 | 448 | 119 | |
GenBank mRNA | BC046086.1 | - | 2 - 3 | 449 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
