Assay Details
Gene Symbol: ttpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199731.2 | NP_956025.2 | 1 - 2 | 263 | 89 | |
GenBank mRNA | BC055617.1 | - | 1 - 2 | 266 | 89 | |
BC165127.1 | - | 1 - 2 | 223 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
