Assay Details
Gene Symbol: tnfaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199799.1 | NP_956093.1 | 4 - 5 | 742 | 126 | |
XM_005157506.4 | XP_005157563.1 | 4 - 5 | 696 | 126 | ||
XM_017359218.2 | XP_017214707.1 | 4 - 5 | 632 | 126 | ||
XM_021481342.1 | XP_021337017.1 | 5 - 6 | 710 | 126 | ||
GenBank mRNA | BC044362.1 | - | 4 - 5 | 742 | 126 | |
BC164349.1 | - | 3 - 4 | 525 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
