Assay Details
Gene Symbol: tnfaip8l2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200374.1 | NP_956668.1 | 2 - 3 | 446 | 95 | |
GenBank mRNA | BC053238.1 | - | 2 - 3 | 446 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
