Assay Details
Gene Symbol: bokb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201185.1 | NP_957479.1 | 3 - 4 | 628 | 89 | |
GenBank mRNA | BC053218.1 | - | 3 - 4 | 628 | 89 | |
BC165297.1 | - | 3 - 4 | 547 | 89 | ||
DQ860157.1 | - | 3 - 4 | 628 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
