Assay Details
Gene Symbol: mmgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205599.1 | NP_991162.1 | 2 - 3 | 190 | 82 | |
GenBank mRNA | AY394962.1 | - | 2 - 3 | 190 | 82 | |
BC076369.1 | - | 2 - 3 | 178 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
