Assay Details
Gene Symbol: mat2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290080.1 | NP_001277009.1 | 1 - 2 | 374 | 74 | |
GenBank mRNA | BC052136.1 | - | 1 - 2 | 373 | 74 | |
JN848834.1 | - | 1 - 2 | 303 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
