Assay Details
Gene Symbol: mat2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290080.1 | NP_001277009.1 | 5 - 6 | 830 | 57 | |
GenBank mRNA | BC052136.1 | - | 5 - 6 | 829 | 57 | |
JN848834.1 | - | 5 - 6 | 759 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
