Assay Details
Gene Symbol: wdr32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212946.1 | NP_998111.2 | 1 - 2 | 585 | 85 | |
GenBank mRNA | BC067591.1 | - | 1 - 2 | 585 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
