Assay Details
Gene Symbol: psmg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213032.1 | NP_998197.1 | 3 - 4 | 372 | 72 | |
GenBank mRNA | BC059607.1 | - | 3 - 4 | 372 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
