Assay Details
Gene Symbol: ppp3r1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004553.1 | NP_001004553.1 | 3 - 4 | 411 | 77 | |
XM_005160126.4 | XP_005160183.1 | 2 - 3 | 274 | 77 | ||
GenBank mRNA | BC081617.1 | - | 3 - 4 | 411 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
