Assay Details
Gene Symbol: gla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006103.2 | NP_001006103.2 | 2 - 3 | 409 | 79 | |
GenBank mRNA | BC083209.1 | - | 2 - 3 | 410 | 80 | |
BC164755.1 | - | 1 - 2 | 158 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
