Assay Details
Gene Symbol: commd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008734.2 | NP_001008734.1 | 1 - 2 | 166 | 71 | |
GenBank mRNA | BC086852.1 | - | 1 - 2 | 169 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
