Assay Details
Gene Symbol: mtmr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014330.2 | NP_001014352.1 | 13 - 14 | 1777 | 102 | |
XM_005161411.4 | XP_005161468.1 | 12 - 13 | 1779 | 102 | ||
GenBank mRNA | BC091956.1 | - | 13 - 14 | 1781 | 102 | |
BC165859.1 | - | 13 - 14 | 1544 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
