Assay Details
Gene Symbol: mtmr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014330.2 | NP_001014352.1 | 8 - 9 | 952 | 66 | |
XM_005161411.4 | XP_005161468.1 | 7 - 8 | 954 | 66 | ||
GenBank mRNA | BC091956.1 | - | 8 - 9 | 956 | 66 | |
BC165859.1 | - | 8 - 9 | 719 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
