Assay Details
Gene Symbol: lta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024821.1 | NP_001019992.1 | 2 - 3 | 171 | 107 | |
XM_009291801.3 | XP_009290076.1 | 3 - 4 | 325 | 107 | ||
GenBank mRNA | AB183468.1 | - | 2 - 3 | 171 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
