Assay Details
Gene Symbol: taf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042744.2 | NP_001036209.1 | 2 - 3 | 432 | 73 | |
GenBank mRNA | BC098542.1 | - | 2 - 3 | 544 | 73 | |
BC116480.1 | - | 2 - 3 | 544 | 73 | ||
BC133110.1 | - | 2 - 3 | 544 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
