Assay Details
Gene Symbol: taf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042744.2 | NP_001036209.1 | 5 - 6 | 2473 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
