Assay Details
Gene Symbol: cyp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131879.2 | NP_571954.1 | 2 - 3 | 924 | 70 | |
GenBank mRNA | AB078927.1 | - | 2 - 3 | 924 | 70 | |
AF210727.2 | - | 2 - 3 | 899 | 70 | ||
AY398333.1 | - | 2 - 3 | 898 | 70 | ||
BC094977.1 | - | 2 - 3 | 924 | 70 | ||
BC165188.1 | - | 1 - 2 | 869 | 70 | ||
CF997194.1 | - | 1 - 2 | 58 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
