Assay Details
Gene Symbol: stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173244.2 | NP_775351.2 | 12 - 13 | 2807 | 62 | |
XM_017353103.2 | XP_017208592.1 | 7 - 8 | 1798 | 62 | ||
GenBank mRNA | AY099525.1 | - | 12 - 13 | 2701 | 62 | |
BC091991.1 | - | 12 - 13 | 2813 | 62 | ||
BC164583.1 | - | 11 - 12 | 2465 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
