Assay Details
Gene Symbol: stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173244.2 | NP_775351.2 | 3 - 4 | 570 | 77 | |
GenBank mRNA | AY099525.1 | - | 3 - 4 | 467 | 77 | |
BC091991.1 | - | 3 - 4 | 576 | 77 | ||
BC164583.1 | - | 2 - 3 | 228 | 77 | ||
EH591838.1 | - | 3 - 4 | 526 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
