Assay Details
Gene Symbol: stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173244.2 | NP_775351.2 | 4 - 5 | 685 | 86 | |
GenBank mRNA | AY099525.1 | - | 4 - 5 | 86 | ||
BC091991.1 | - | 4 - 5 | 691 | 86 | ||
BC164583.1 | - | 3 - 4 | 343 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
