Assay Details
Gene Symbol: stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173244.2 | NP_775351.2 | 6 - 7 | 1106 | 60 | |
GenBank mRNA | AY099525.1 | - | 6 - 7 | 1003 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
