Assay Details
Gene Symbol: stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173244.2 | NP_775351.2 | 7 - 8 | 1195 | 99 | |
XM_017353103.2 | XP_017208592.1 | 2 - 3 | 186 | 99 | ||
GenBank mRNA | AY099525.1 | - | 7 - 8 | 1092 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
