Assay Details
Gene Symbol: elovl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199532.1 | NP_955826.1 | 3 - 4 | 498 | 80 | |
GenBank mRNA | BC059459.1 | - | 3 - 4 | 498 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
