Assay Details
Gene Symbol: det1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199670.1 | NP_955964.1 | 3 - 4 | 1322 | 100 | |
XM_009297856.3 | XP_009296131.1 | 3 - 4 | 1571 | 100 | ||
GenBank mRNA | BC055677.1 | - | 3 - 4 | 1322 | 100 | |
BC076520.1 | - | 3 - 4 | 1292 | 100 | ||
BC164661.1 | - | 2 - 3 | 1159 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
