Assay Details
Gene Symbol: det1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199670.1 | NP_955964.1 | 4 - 5 | 1507 | 76 | |
XM_009297856.3 | XP_009296131.1 | 4 - 5 | 1756 | 76 | ||
GenBank mRNA | BC055677.1 | - | 4 - 5 | 1507 | 76 | |
BC076520.1 | - | 4 - 5 | 1477 | 76 | ||
BC164661.1 | - | 3 - 4 | 1344 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
