Assay Details
Gene Symbol: chmp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200014.1 | NP_956308.1 | 7 - 8 | 689 | 63 | |
XM_009301289.3 | XP_009299564.1 | 7 - 8 | 726 | 63 | ||
GenBank mRNA | AY398326.1 | - | 7 - 8 | 695 | 63 | |
BC045934.1 | - | 7 - 8 | 689 | 63 | ||
BC065462.1 | - | 7 - 8 | 672 | 63 | ||
BC067569.1 | - | 7 - 8 | 665 | 63 | ||
BC153497.1 | - | 7 - 8 | 693 | 63 | ||
BC165138.1 | - | 7 - 8 | 593 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
