Assay Details
Gene Symbol: ggps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291760.1 | NP_001278689.1 | 3 - 4 | 453 | 126 | |
XM_017358708.2 | XP_017214197.1 | 3 - 4 | 499 | 126 | ||
GenBank mRNA | BC052115.1 | - | 4 - 5 | 356 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
