Assay Details
Gene Symbol: chm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203461.1 | NP_982286.1 | 12 - 13 | 1484 | 67 | |
GenBank mRNA | AY500891.1 | - | 12 - 13 | 1484 | 67 | |
BC096773.1 | - | 12 - 13 | 1517 | 67 | ||
BC154207.1 | - | 12 - 13 | 1498 | 67 | ||
BC164561.1 | - | 12 - 13 | 1500 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
