Assay Details
Gene Symbol: chm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203461.1 | NP_982286.1 | 9 - 10 | 1213 | 68 | |
GenBank mRNA | AY500891.1 | - | 9 - 10 | 1213 | 68 | |
BC154207.1 | - | 9 - 10 | 1227 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
