Assay Details
Gene Symbol: cacna1da | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203484.1 | NP_982351.1 | 14 - 15 | 2069 | 68 | |
XM_021479576.1 | XP_021335251.1 | 14 - 15 | 2257 | 68 | ||
XM_021479577.1 | XP_021335252.1 | 14 - 15 | 2257 | 68 | ||
XM_021479578.1 | XP_021335253.1 | 14 - 15 | 2257 | 68 | ||
XM_021479579.1 | XP_021335254.1 | 14 - 15 | 2257 | 68 | ||
XM_021479580.1 | XP_021335255.1 | 14 - 15 | 2256 | 68 | ||
XM_021479581.1 | XP_021335256.1 | 13 - 14 | 2181 | 68 | ||
XM_021479582.1 | XP_021335257.1 | 12 - 13 | 1876 | 68 | ||
XM_021479583.1 | XP_021335258.1 | 14 - 15 | 2257 | 68 | ||
XM_021479584.1 | XP_021335259.1 | 13 - 14 | 2181 | 68 | ||
GenBank mRNA | AY528224.1 | - | 14 - 15 | 2069 | 68 | |
BC163679.1 | - | 12 - 13 | 1876 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
